5-Iod-α,α,α,α',α',α'-Hexafluor-m-xylol

Lieferant: FLUOROCHEM

Synonyme: 3,5-Bis(trifluormethyl)iodbenzol, 1-Iod-3,5-bis(trifluormethyl)benzol, 1,3-Bis(trifluormethyl)-5-iodbenzol, 5-Iod-α,α,α,β,β,β-Hexafluor-m-xylol

006029-25G 006029-10G
FLUO006029-25GEA 110 EUR
FLUO006029-25G FLUO006029-10G
5-Iod-α,α,α,α',α',α'-Hexafluor-m-xylol
5-Iod-α,α,α,α',α',α'-Hexafluor-m-xylol
Formel: C₈H₃F₆I
Molecular Weight: 340,01 g/mol
Siedepunkt: 171…172 °C (1013 hPa)
Dichte: 1,91 g/cm³ (20 °C)
MDL: MFCD00040837
CAS-Nummer: 328-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR