2,4-Dibrom-5-fluoranisol

Lieferant: COMBI-BLOCKS

Synonyme: 1,3-Dibrom-4-fluor-6-methoxybenzol

OT-0975-5G OT-0975-1G
COBBOT-0975-5GEA 451 EUR
COBBOT-0975-5G COBBOT-0975-1G
2,4-Dibrom-5-fluoranisol
2,4-Dibrom-5-fluoranisol
Formel: C₇H₅Br₂FO
Molecular Weight: 283.92 g/mol
MDL: MFCD12913976
CAS-Nummer: 861928-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR