2,6-Dibrom-4-fluoranisol

Lieferant: COMBI-BLOCKS

OT-0812-5G OT-0812-1G
COBBOT-0812-5GEA 229 EUR
COBBOT-0812-5G COBBOT-0812-1G
2,6-Dibrom-4-fluoranisol
2,6-Dibrom-4-fluoranisol
Formel: C₇H₅Br₂FO
Molecular Weight: 283.92 g/mol
MDL: MFCD00042231
CAS-Nummer: 443-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR