3,5-Dibromanisol

Lieferant: COMBI-BLOCKS

Synonyme: 1,3-Dibromo-5-methoxybenzene

OT-0386-1G OT-0386-25G OT-0386-5G
COBBOT-0386-1GEA 0 EUR
COBBOT-0386-1G COBBOT-0386-25G COBBOT-0386-5G
3,5-Dibromanisol
3,5-Dibromanisol
Formel: C₇H₆Br₂O
Molecular Weight: 265,93 g/mol
MDL: MFCD02258848
CAS-Nummer: 74137-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR