2',4'-Dichlor-5'-fluoracetophenon

Lieferant: Thermo Scientific

Synonyme: 1-(2,4-Dichloro-5-fluorophenyl)ethanone

BTB15028.1 BTB15028.10 BTB15028.50
MAYBBTB15028.1EA 0 EUR
MAYBBTB15028.1 MAYBBTB15028.10 MAYBBTB15028.50
2',4'-Dichlor-5'-fluoracetophenon
2',4'-Dichlor-5'-fluoracetophenon
Formel: C₈H₅Cl₂FO
Molecular Weight: 207,03 g/mol
MDL: MFCD00077499
CAS-Nummer: 704-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR