1,3-Dichlor-4-fluor-6-nitrobenzol

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dichlor-5-fluornitrobenzol

008279-1G 008279-5G
FLUO008279-1GEA 149 EUR
FLUO008279-1G FLUO008279-5G
1,3-Dichlor-4-fluor-6-nitrobenzol
1,3-Dichloro-4-fluoro-6-nitrobenzene
Formel: C₆H₂Cl₂FNO₂
Molecular Weight: 209.99 g/mol
MDL: MFCD00115003
CAS-Nummer: 2105-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR