3,5-Difluorphenol
Lieferant: COMBI-BLOCKS
OT-0889-5G
OT-0889-25G
OT-0889-100G
COBBOT-0889-5GEA
50.3
EUR
COBBOT-0889-5G
COBBOT-0889-25G
COBBOT-0889-100G
3,5-Difluorphenol
3,5-Difluorphenol
Formel:
C₆H₄F₂O Molecular Weight: 130.09 g/mol Siedepunkt: 65…68 °C (15 mmHg) Schmelzpunkt: 55 Dichte: 1.566 g/cm³ (20 °C) |
MDL:
MFCD00002255 CAS-Nummer: 2713-34-0 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...