3,5-Difluortoluol

Lieferant: COMBI-BLOCKS

Synonyme: 1,3-Difluor-5-methylbenzol

OS-7677-25G OS-7677-5G OS-7677-1G
COBBOS-7677-25GEA 410 EUR
COBBOS-7677-25G COBBOS-7677-5G COBBOS-7677-1G
3,5-Difluortoluol
3,5-Difluortoluol
Formel: C₇H₆F₂
Molecular Weight: 128,12 g/mol
MDL: MFCD00797323
CAS-Nummer: 117358-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR