2',4'-Dihydroxy-3'-methylacetophenon

Lieferant: Thermo Scientific

Synonyme: 1-(2,4-Dihydroxy-3-methylphenyl)ethan-1-on

RH00145.1 RH00145.10 RH00145.50
MAYBRH00145.1EA 0 EUR
MAYBRH00145.1 MAYBRH00145.10 MAYBRH00145.50
2',4'-Dihydroxy-3'-methylacetophenon
2',4'-Dihydroxy-3'-methylacetophenon
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
MDL: MFCD00010817
CAS-Nummer: 10139-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR