1,3-Dimethoxy-2-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1236-5G AN-1236-1G
COBBAN-1236-5GEA 569 EUR
COBBAN-1236-5G COBBAN-1236-1G
1,3-Dimethoxy-2-nitrobenzol
1,3-Dimethoxy-2-nitrobenzol
Formel: C₈H₉NO₄
Molecular Weight: 183.16 g/mol
MDL: MFCD00474587
CAS-Nummer: 6665-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR