2',4'-Dimethylacetophenon

Lieferant: Thermo Scientific

Synonyme: 1-(2,4-Dimethylphenyl)ethan-1-on

SB00787.1 SB00787.10 SB00787.50
MAYBSB00787.1EA 0 EUR
MAYBSB00787.1 MAYBSB00787.10 MAYBSB00787.50
2',4'-Dimethylacetophenon
2',4'-Dimethylacetophenon
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
Siedepunkt: 70…72 °C (0,5 mmHg)
Dichte: 1,0121 g/cm³ (15 °C)
MDL: MFCD00003571
CAS-Nummer: 89-74-7
EINECS: 201-935-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR