3,5-Dinitrobenzylalkohol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H32669.06
H32669.14
H32669.22
H32669.06EA
60.6
EUR
H32669.06
H32669.14
H32669.22
3,5-Dinitrobenzylalkohol ≥98%
3,5-Dinitrobenzylalkohol
Purity: 98%
CAS No.: 71022-43-0
Molecular Formula: C7H6N2O5
Formula Weight: 198.13
Melting Point: 88-91°
Beilstein Reference: 2054388
Formel:
C₇H₆N₂O₅ Molecular Weight: 198,14 g/mol Schmelzpunkt: 88…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007235 CAS-Nummer: 71022-43-0 EINECS: 275-131-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...