1,4-Diaminobutan Dihydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,4-Butandiamin Dihydrochlorid, Tetramethylendiammoniumdichlorid, Tetramethylendiamin Dihydrochlorid, Putrescin Dihydrochlorid
M-Clarity™ Qualitätsstufe = MQ400
32810-100G
32810-25G
SIAL32810-100GEA
202
EUR
SIAL32810-100G
SIAL32810-25G
1,4-Diaminobutan Dihydrochlorid, Sigma-Aldrich®
1,4-Diaminobutan Dihydrochlorid
Formel:
C₄H₁₄Cl₂N₂ Molecular Weight: 161,07 g/mol Dichte: 1,31 g/cm³ (20 °C) |
MDL:
MFCD00012526 CAS-Nummer: 333-93-7 EINECS: 206-375-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...