2,5-Dibromphenol
Lieferant: COMBI-BLOCKS
Synonyme:
1,3-Dibrom-4-hydroxybenzol
OT-1198-1G
OT-1198-25G
OT-1198-5G
COBBOT-1198-1GEA
73.5
EUR
COBBOT-1198-1G
COBBOT-1198-25G
COBBOT-1198-5G
2,5-Dibromphenol
2,5-Dibromphenol
Formel:
C₆H₄Br₂O Molecular Weight: 251,91 g/mol |
MDL:
MFCD01851373 CAS-Nummer: 28165-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...