2',5'-Dimethylacetophenon
Lieferant: CHEMPUR
Synonyme:
1-(2,5-Dimethylphenyl)ethan-1-on
BTB06231.250
BTB06231.1
CHMPBTB06231.250EA
0
EUR
CHMPBTB06231.250
MAYBBTB06231.1
2',5'-Dimethylacetophenon
2',5'-Dimethylacetophenon
Formel:
C₁₀H₁₂O Molecular Weight: 148.2 g/mol Siedepunkt: 231 Schmelzpunkt: –18 °C Dichte: 1.0154 g/cm³ (0 °C) |
MDL:
MFCD00014988 CAS-Nummer: 2142-73-6 EINECS: 218-399-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...