1,6-Diaminohexan Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,6-Hexandiamin Dihydrochlorid, Hexamethylendiammoniumdichlorid, Hexamethylendiamin Dihydrochlorid

M-Clarity™ Qualitätsstufe = MQ200

247731-100G 247731-25G
SIAL247731-100GEA 143 EUR
SIAL247731-100G SIAL247731-25G
1,6-Diaminohexan Dihydrochlorid, Sigma-Aldrich®
1,6-Diaminohexan Dihydrochlorid
Formel: C₆H₁₈Cl₂N₂
Molecular Weight: 189,13 g/mol
Schmelzpunkt: 258 °C
Dichte: 1,216 g/cm³ (20 °C)
MDL: MFCD00012528
CAS-Nummer: 6055-52-3
EINECS: 227-977-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR