2,4-Difluor-6-nitroanilin

Lieferant: COMBI-BLOCKS

HC-2914-5G HC-2914-25G
COBBHC-2914-5GEA 37.5 EUR
COBBHC-2914-5G COBBHC-2914-25G
2,4-Difluor-6-nitroanilin
2,4-Difluor-6-nitroanilin
Formel: C₆H₄F₂N₂O₂
Molecular Weight: 174,11 g/mol
MDL: MFCD00034058
CAS-Nummer: 364-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR