2,6-Dibrom-p-toluidin
Lieferant: COMBI-BLOCKS
Synonyme:
2,6-Dibrom-4-methylanilin, 4-Amino-3,5-dibromtoluol
AN-1556-25G
AN-1556-100G
COBBAN-1556-25GEA
38.5
EUR
COBBAN-1556-25G
COBBAN-1556-100G
2,6-Dibrom-p-toluidin
2,6-Dibrom-p-toluidin
Formel:
C₇H₇Br₂N Molecular Weight: 264,95 g/mol Siedepunkt: 296 °C (1013 hPa) Schmelzpunkt: 79 °C |
MDL:
MFCD00007641 CAS-Nummer: 6968-24-7 EINECS: 230-182-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...