2-Brom-4-(trifluormethoxy)anilin
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-4-trifluoromethoxyaniline
AN-1178-5G
AN-1178-25G
AN-1178-100G
COBBAN-1178-5GEA
25.5
EUR
COBBAN-1178-5G
COBBAN-1178-25G
COBBAN-1178-100G
2-Brom-4-(trifluormethoxy)anilin
2-Brom-4-(trifluormethoxy)anilin
Formel:
C₇H₅BrF₃NO Molecular Weight: 256.02 g/mol |
MDL:
MFCD00203478 CAS-Nummer: 175278-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...