2-Brom-4-chlor-6-fluoranilin

Lieferant: COMBI-BLOCKS

AN-2570-100G AN-2570-5G AN-2570-25G
COBBAN-2570-100GEA 410 EUR
COBBAN-2570-100G COBBAN-2570-5G COBBAN-2570-25G
2-Brom-4-chlor-6-fluoranilin
2-Brom-4-chlor-6-fluoranilin
Formel: C₆H₄BrClFN
Molecular Weight: 224,46 g/mol
MDL: MFCD00070746
CAS-Nummer: 195191-47-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR