2-Brom-6-chlor-4-fluoranilin

Lieferant: COMBI-BLOCKS

OT-0399-5G OT-0399-25G OT-0399-1G
COBBOT-0399-5GEA 63.4 EUR
COBBOT-0399-5G COBBOT-0399-25G COBBOT-0399-1G
2-Brom-6-chlor-4-fluoranilin
2-Brom-6-chlor-4-fluoranilin
Formel: C₆H₄BrClFN
Molecular Weight: 224,46 g/mol
Siedepunkt: 260 °C (1013 hPa)
Schmelzpunkt: 55…56 °C
MDL: MFCD00051707
CAS-Nummer: 201849-14-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR