2-Amino-3-brom-5-nitrobenzonitril

Lieferant: COMBI-BLOCKS

AN-1129-5G AN-1129-25G AN-1129-100G
COBBAN-1129-5GEA 37.5 EUR
COBBAN-1129-5G COBBAN-1129-25G COBBAN-1129-100G
2-Amino-3-brom-5-nitrobenzonitril
2-Amino-3-brom-5-nitrobenzonitril
Formel: C₇H₄BrN₃O₂
Molecular Weight: 242.03 g/mol
Schmelzpunkt: 233…235 °C
Dichte: 1.949
MDL: MFCD00054185
CAS-Nummer: 17601-94-4
EINECS: 241-574-4
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR