2-Chlor-4-(trifluormethyl)anilin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
4-Amino-3-chlor-α,α,α-trifluortoluol, 2-Chlor-α,α,α-trifluor-p-toluidin
AN-1491-25G
AN-1491-100G
COBBAN-1491-25GEA
69.2
EUR
COBBAN-1491-25G
COBBAN-1491-100G
2-Chlor-4-(trifluormethyl)anilin
2-Chlor-4-(trifluormethyl)anilin
Formel:
C₇H₅ClF₃N Molecular Weight: 195.57 g/mol Siedepunkt: 89 °C (0.5 mmHg) |
MDL:
MFCD00042563 CAS-Nummer: 39885-50-2 EINECS: 254-674-8 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...