5-(Trifluormethyl)-o-anisidin
Lieferant: COMBI-BLOCKS
Synonyme:
3-Amino-4-methoxybenzotrifluorid, 5-(Trifluormethyl)-2-methoxyanilin, 2-Methoxy-5-(trifluormethyl)anilin, 2-Amino-4-(trifluormethyl)anisol, 5-(Trifluormethyl)-2-anisidin
HC-6980-5G
HC-6980-25G
HC-6980-100G
COBBHC-6980-5GEA
30.8
EUR
COBBHC-6980-5G
COBBHC-6980-25G
COBBHC-6980-100G
5-(Trifluormethyl)-o-anisidin
5-(Trifluormethyl)-o-anisidin
Formel:
C₈H₈F₃NO Molecular Weight: 191.15 g/mol Siedepunkt: 230 Schmelzpunkt: 59 |
MDL:
MFCD00042486 CAS-Nummer: 349-65-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...