3-Brom-4-fluoranilin
Lieferant: COMBI-BLOCKS
OS-7480-25G
OS-7480-5G
OS-7480-100G
COBBOS-7480-25GEA
50.8
EUR
COBBOS-7480-25G
COBBOS-7480-5G
COBBOS-7480-100G
3-Brom-4-fluoranilin
3-Brom-4-fluoranilin
Formel:
C₆H₅BrFN Molecular Weight: 190.01 g/mol |
MDL:
MFCD03788559 CAS-Nummer: 656-64-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...