2-Amino-4-bromphenol

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-hydroxyanilin

HC-6973-5G HC-6973-25G HC-6973-1G
COBBHC-6973-5GEA 30.8 EUR
COBBHC-6973-5G COBBHC-6973-25G COBBHC-6973-1G
2-Amino-4-bromphenol
2-Amino-4-bromphenol
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD00235171
CAS-Nummer: 40925-68-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR