3-Chlor-2,6-dibrom-4-fluoranilin

Lieferant: FLUOROCHEM

Synonyme: 2,6-Dibromo-3-chloro-4-fluoroaniline, 1-Amino-3-chlor-2,6-dibrom-4-fluorbenzol

004386-1G 004386-25G 004386-5G
FLUO004386-1GEA 31.1 EUR
FLUO004386-1G FLUO004386-25G FLUO004386-5G
3-Chlor-2,6-dibrom-4-fluoranilin
3-Chlor-2,6-dibrom-4-fluoranilin
Formel: C₆H₃Br₂ClFN
Molecular Weight: 303,36 g/mol
Siedepunkt: 302 °C (1013 hPa)
Schmelzpunkt: 97…99 °C
MDL: MFCD00042199
CAS-Nummer: 175135-09-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR