3-Chlor-2-nitroanilin

Lieferant: COMBI-BLOCKS
Gefahr

OS-7452-5G OS-7452-25G OS-7452-1G
COBBOS-7452-5GEA 76.5 EUR
COBBOS-7452-5G COBBOS-7452-25G COBBOS-7452-1G
3-Chlor-2-nitroanilin
3-Chlor-2-nitroanilin
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01317803
CAS-Nummer: 59483-54-4
EINECS: 261-782-9
UN: 2237
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR