3-Chlor-p-anisidin
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chlor-4-anisidin, 3-Chlor-4-methoxyanilin, 4-Amino-2-chloranisol
AN-1485-5G
AN-1485-25G
AN-1485-100G
COBBAN-1485-5GEA
37.5
EUR
COBBAN-1485-5G
COBBAN-1485-25G
COBBAN-1485-100G
3-Chlor-p-anisidin
3-Chlor-p-anisidin
Formel:
C₇H₈ClNO Molecular Weight: 157,6 g/mol Siedepunkt: 253 °C (1013 hPa) Schmelzpunkt: 82 °C |
MDL:
MFCD00007771 CAS-Nummer: 5345-54-0 EINECS: 226-298-4 UN: 2233 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...