3-Cyan-4-fluoranilin Hydrochlorid

Lieferant: COMBI-BLOCKS

AN-1041-25G AN-1041-5G
COBBAN-1041-25GEA 0 EUR
COBBAN-1041-25G COBBAN-1041-5G
3-Cyan-4-fluoranilin Hydrochlorid
3-Cyan-4-fluoranilin Hydrochlorid
Formel: C₇H₆ClFN₂
MDL: MFCD01632195
CAS-Nummer: 801316-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR