3-Fluor-p-anisidin
Lieferant: COMBI-BLOCKS
Synonyme:
4-Amino-2-fluoranisol, 3-Fluor-4-methoxyanilin, 3-Fluor-4-anisidin
AN-1094-5G
AN-1094-25G
COBBAN-1094-5GEA
31
EUR
COBBAN-1094-5G
COBBAN-1094-25G
3-Fluor-p-anisidin
3-Fluor-p-anisidin
Formel:
C₇H₈FNO |
MDL:
MFCD00075040 CAS-Nummer: 366-99-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...