5-Fluor-o-anisidin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Fluor-2-anisidin, 2-Amino-4-fluoranisol, 5-Fluor-2-methoxyanilin
AN-1152-25G
AN-1152-5G
AN-1152-1G
COBBAN-1152-25GEA
269
EUR
COBBAN-1152-25G
COBBAN-1152-5G
COBBAN-1152-1G
5-Fluor-o-anisidin
5-Fluor-o-anisidin
Formel:
C₇H₈FNO Molecular Weight: 141,15 g/mol |
MDL:
MFCD00077537 CAS-Nummer: 1978-39-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...