5-Amino-o-kresol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Hydroxy-4-methylanilin, 4-Amino-2-hydroxytoluol, 1-Amino-3-hydroxy-4-methylbenzol, 5-Amino-2-methylphenol

AN-1203-25G AN-1203-100G
COBBAN-1203-25GEA 47.3 EUR
COBBAN-1203-25G COBBAN-1203-100G
5-Amino-o-kresol
5-Amino-o-kresol
Formel: C₇H₉NO
Molecular Weight: 123,15 g/mol
Siedepunkt: 259 °C (1013 hPa)
Schmelzpunkt: 162 °C
MDL: MFCD00043922
CAS-Nummer: 2835-95-2
EINECS: 220-618-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR