4-Brom-2-(trifluormethoxy)anilin
Lieferant: COMBI-BLOCKS
OS-7489-5G
OS-7489-25G
OS-7489-100G
COBBOS-7489-5GEA
22
EUR
COBBOS-7489-5G
COBBOS-7489-25G
COBBOS-7489-100G
4-Brom-2-(trifluormethoxy)anilin
4-Brom-2-(trifluormethoxy)anilin
Formel:
C₇H₅BrF₃NO Molecular Weight: 256.02 g/mol |
MDL:
MFCD00179338 CAS-Nummer: 175278-09-8 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...