4-Brom-2-chloranilin
Lieferant: COMBI-BLOCKS
|
Achtung
|
OS-7470-5G
OS-7470-25G
OS-7470-100G
COBBOS-7470-5GEA
21.6
EUR
COBBOS-7470-5G
COBBOS-7470-25G
COBBOS-7470-100G
4-Brom-2-chloranilin
4-Brom-2-chloranilin
Formel:
C₆H₅BrClN Molecular Weight: 206.47 g/mol Siedepunkt: 264 Schmelzpunkt: 71 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007660 CAS-Nummer: 38762-41-3 EINECS: 254-118-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...