4-Brom-3-chlor-2-fluoranilin

Lieferant: COMBI-BLOCKS

AN-1104-100G AN-1104-25G
COBBAN-1104-100GEA 398 EUR
COBBAN-1104-100G COBBAN-1104-25G
4-Brom-3-chlor-2-fluoranilin
4-Brom-3-chlor-2-fluoranilin
Formel: C₆H₄BrClFN
Molecular Weight: 224.46 g/mol
MDL: MFCD04039280
CAS-Nummer: 115843-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR