4-Brom-1-naphthylamin

Lieferant: COMBI-BLOCKS

Synonyme: 1-Amino-4-bromnaphthalin

AN-1105-100G AN-1105-5G AN-1105-25G
COBBAN-1105-100GEA 385 EUR
COBBAN-1105-100G COBBAN-1105-5G COBBAN-1105-25G
4-Brom-1-naphthylamin
4-Brom-1-naphthylamin
Formel: C₁₀H₈BrN
Molecular Weight: 222.08 g/mol
MDL: MFCD00004023
CAS-Nummer: 2298-07-9
EINECS: 218-944-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR