2-Amino-2',5-dichlorbenzophenon 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (2-Amino-5-chlorophenyl)(2-chlorophenyl)methanone

B20360.06 B20360.09 B20360.18
B20360.06EA 44.2 EUR
B20360.06 B20360.09 B20360.18
2-Amino-2',5-dichlorbenzophenon 99%
2-Amino-2',5-dichlorbenzophenon

Purity: 99% CAS No.: 2958-36-3 Molecular Formula: C13H9Cl2NO Formula Weight: 266.13 Odour: Odourless Melting Point: 87-89° Beilstein Reference: 652774 MDL No.: MFCD00007840

Formel: C₁₃H₉Cl₂NO
Molecular Weight: 266.13 g/mol
Siedepunkt: 385
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007840
CAS-Nummer: 2958-36-3
EINECS: 220-985-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR