2-Amino-5-chlor-2'-fluorbenzophenon

Lieferant: CHEMPUR

Synonyme: (2-Amino-5-chlorophenyl)(2-fluorophenyl)methanone, 2-Amino-5-chloro-2′-fluorobenzophenone

FL3839.25 FL3839.100
CHMPFL3839.25EA 0 EUR
CHMPFL3839.25 CHMPFL3839.100
2-Amino-5-chlor-2'-fluorbenzophenon
2-Amino-5-chlor-2'-fluorbenzophenon
Formel: C₁₃H₉ClFNO
Molecular Weight: 249.67 g/mol
Siedepunkt: 364
Schmelzpunkt: 94…95 °C
MDL: MFCD00038381
CAS-Nummer: 784-38-3
EINECS: 212-316-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR