2,6-Dibrom-4-chloranilin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
4-Chloro-2,6-dibromoaniline, 1-Amino-4-chloro-2,6-dibromobenzene
OT-0623-100G
OT-0623-25G
OT-0623-5G
COBBOT-0623-100GEA
205
EUR
COBBOT-0623-100G
COBBOT-0623-25G
COBBOT-0623-5G
2,6-Dibrom-4-chloranilin
2,6-Dibrom-4-chloranilin
Formel:
C₆H₄Br₂ClN Molecular Weight: 285,37 g/mol Siedepunkt: 301 °C (1013 hPa) Schmelzpunkt: 96 °C |
MDL:
MFCD00051750 CAS-Nummer: 874-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...