4-Chlor-m-anisidin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Amino-2-chloranisol, 4-Chlor-3-anisidin, 4-Chlor-3-methoxyanilin

AN-1266-5G AN-1266-25G AN-1266-1G
COBBAN-1266-5GEA 88.6 EUR
COBBAN-1266-5G COBBAN-1266-25G COBBAN-1266-1G
4-Chlor-m-anisidin
4-Chlor-m-anisidin
Formel: C₇H₈ClNO
Molecular Weight: 157.6 g/mol
MDL: MFCD00672967
CAS-Nummer: 13726-14-2
UN: 2233
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR