4-Fluor-m-anisidin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluor-3-anisidin, 5-Amino-2-fluoranisol, 4-Fluor-3-methoxyanilin

AN-1070-5G AN-1070-25G AN-1070-1G
COBBAN-1070-5GEA 63.4 EUR
COBBAN-1070-5G COBBAN-1070-25G COBBAN-1070-1G
4-Fluor-m-anisidin
4-Fluor-m-anisidin
Formel: C₇H₈FNO
Molecular Weight: 141,15 g/mol
MDL: MFCD00665789
CAS-Nummer: 64465-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR