2-(2-Bromphenyl)ethanol

Lieferant: COMBI-BLOCKS

OT-0499-5G OT-0499-25G OT-0499-1G
COBBOT-0499-5GEA 38.2 EUR
COBBOT-0499-5G COBBOT-0499-25G COBBOT-0499-1G
2-(2-Bromphenyl)ethanol
2-(2-Bromphenyl)ethanol
Formel: C₈H₉BrO
Molecular Weight: 201.06 g/mol
MDL: MFCD00093566
CAS-Nummer: 1074-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR