2-Brombenzylalkohol
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Bromophenyl)methanol
OS-7635-5G
OS-7635-25G
OS-7635-100G
COBBOS-7635-5GEA
25.5
EUR
COBBOS-7635-5G
COBBOS-7635-25G
COBBOS-7635-100G
2-Brombenzylalkohol
2-Brombenzylalkohol
Formel:
C₇H₇BrO Molecular Weight: 187,04 g/mol Siedepunkt: 255 °C (1013 hPa) Schmelzpunkt: 80…82 °C |
MDL:
MFCD00004600 CAS-Nummer: 18982-54-2 EINECS: 242-719-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...