1-Brom-2,3,5-trichlorbenzol
Lieferant: COMBI-BLOCKS
OS-7628-5G
OS-7628-25G
OS-7628-1G
COBBOS-7628-5GEA
88.6
EUR
COBBOS-7628-5G
COBBOS-7628-25G
COBBOS-7628-1G
1-Brom-2,3,5-trichlorbenzol
1-Bromo-2,3,5-trichlorobenzene
Formel:
C₆H₂BrCl₃ Molecular Weight: 260,34 g/mol |
MDL:
MFCD00070734 CAS-Nummer: 81067-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...