1-Brom-2,3-difluorbenzol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2,3-Difluorobromobenzene
OS-7592-5G
OS-7592-25G
OS-7592-100G
COBBOS-7592-5GEA
26
EUR
COBBOS-7592-5G
COBBOS-7592-25G
COBBOS-7592-100G
1-Brom-2,3-difluorbenzol
1-Bromo-2,3-difluorobenzene
Formel:
C₆H₃BrF₂ Molecular Weight: 192,99 g/mol Siedepunkt: 234 °C (765 mmHg) Dichte: 1,72 g/cm³ (20 °C) |
MDL:
MFCD00061136 CAS-Nummer: 38573-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...