1-Brom-2,4-difluor-5-nitrobenzol

Lieferant: BIONET RESEARCH

Synonyme: 5-Brom-2,4-difluornitrobenzol

7X-0836.500 7X-0836.1 7X-0836.5 7X-0836.10
BIOT7X-0836.500EA 0 EUR
BIOT7X-0836.500 BIOT7X-0836.1 BIOT7X-0836.5 BIOT7X-0836.10
1-Brom-2,4-difluor-5-nitrobenzol
1-Bromo-2,4-difluoro-5-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Molecular Weight: 237.99 g/mol
MDL: MFCD00129166
CAS-Nummer: 345-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR