4-Brom-2,5-difluoranisol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 1-Bromo-2,5-difluoro-4-methoxybenzene

OT-0928-5G OT-0928-25G
COBBOT-0928-5GEA 71.7 EUR
COBBOT-0928-5G COBBOT-0928-25G
4-Brom-2,5-difluoranisol
4-Brom-2,5-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD00143258
CAS-Nummer: 202865-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR