1-Brom-2,5-difluor-4-nitrobenzol

Lieferant: CHEMPUR

Synonyme: 4-Brom-2,5-difluornitrobenzol

HAN00007.250 HAN00007.1
CHMPHAN00007.250EA 0 EUR
CHMPHAN00007.250 MAYBHAN00007.1
1-Brom-2,5-difluor-4-nitrobenzol
1-Bromo-2,5-difluoro-4-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Molecular Weight: 237,99 g/mol
Schmelzpunkt: 55…57 °C
MDL: MFCD00042184
CAS-Nummer: 167415-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR