4-Brom-3,5-difluoranisol
Lieferant: COMBI-BLOCKS
|
Achtung
|
OT-0927-25G
OT-0927-1G
OT-0927-5G
COBBOT-0927-25GEA
247
EUR
COBBOT-0927-25G
COBBOT-0927-1G
COBBOT-0927-5G
4-Brom-3,5-difluoranisol
4-Brom-3,5-difluoranisol
Formel:
C₇H₅BrF₂O Molecular Weight: 223,02 g/mol |
MDL:
MFCD00143268 CAS-Nummer: 202865-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...